2,3,4,5,6-Pentafluoro-DL-phenylalanine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-(pentafluorophenyl)propionic acid, 2-Amino-3-(pentafluorophenyl)propanoic acid
PC0781-250MG
APOSPC0781-250MGEA
125
EUR
APOSPC0781-250MG
2,3,4,5,6-Pentafluoro-DL-phenylalanine 97%
2,3,4,5,6-Pentafluoro-DL-phenylalanine
Formel:
C₉H₆F₅NO₂ Molecular Weight: 255,141 g/mol Schmelzpunkt: 254…258 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054668 CAS-Nummer: 3321-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...