3-Bromphenylsulfurpentafluorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3-(pentafluorosulphanyl)benzene
PC0782-1G
PC0782-25G
PC0782-5G
APOSPC0782-1GEA
348
EUR
APOSPC0782-1G
APOSPC0782-25G
APOSPC0782-5G
3-Bromphenylsulfurpentafluorid 95%
3-Bromphenylsulfurpentafluorid
Formel:
C₆H₄BrF₅S Molecular Weight: 283.06 g/mol Dichte: 1.831 g/cm³ (25 °C) Flammpunkt: 87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425927 CAS-Nummer: 672-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...