2-Fluor-4-(trifluormethyl)zimtsäure 98%

Lieferant: Apollo Scientific

PC0783-10G PC0783-5G PC0783-1G
APOSPC0783-10GEA 0 EUR
APOSPC0783-10G APOSPC0783-5G APOSPC0783-1G
2-Fluor-4-(trifluormethyl)zimtsäure 98%
2-Fluor-4-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00236292
CAS-Nummer: 262608-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR