5-Nitro-2-[3-(trifluoromethyl)phenoxy]benzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC0786-10G PC0786-5G
APOSPC0786-10GEA 200 EUR
APOSPC0786-10G APOSPC0786-5G
5-Nitro-2-[3-(trifluoromethyl)phenoxy]benzaldehyde 97%
5-Nitro-2-[3-(trifluoromethyl)phenoxy]benzaldehyde
Formel: C₁₄H₈F₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03001331
CAS-Nummer: 217186-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR