3-Iodphenylsulfurpentafluorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Iodo-3-(pentafluorosulphanyl)benzene
PC0787-1G
APOSPC0787-1GEA
212
EUR
APOSPC0787-1G
3-Iodphenylsulfurpentafluorid
3-Iodphenylsulfurpentafluorid
Formel:
C₆H₄F₅IS Molecular Weight: 330.575 g/mol Siedepunkt: 57 °C/0.75mmHg Dichte: 2.06 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD03788513 CAS-Nummer: 286947-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...