2,2,3,3-Tetrafluor-1,4-benzodioxan

Lieferant: Apollo Scientific
Achtung

PC0789-1G
APOSPC0789-1GEA 261 EUR
APOSPC0789-1G
2,2,3,3-Tetrafluor-1,4-benzodioxan
2,2,3,3-Tetrafluor-1,4-benzodioxan
Formel: C₈H₄F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00236308
CAS-Nummer: 94767-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR