4-(Pentafluorthio)phenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenylsulphur pentafluoride
PC0794-1G
PC0794-5G
APOSPC0794-1GEA
551
EUR
APOSPC0794-1G
APOSPC0794-5G
4-(Pentafluorthio)phenol 97%
4-(Pentafluorthio)phenol
Formel:
C₆H₅F₅OS Schmelzpunkt: 104…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788516 CAS-Nummer: 774-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...