3-(Pentafluorthio)benzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

PC0796-1G
APOSPC0796-1GEA 0 EUR
APOSPC0796-1G
3-(Pentafluorthio)benzaldehyd 97%
3-(Pentafluorothio)benzaldehyde
Formel: C₇H₅F₅OS
Lagertemperatur: Raumtemperatur
MDL: MFCD03788517
CAS-Nummer: 401892-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR