4-(Pentafluorthio)benzaldehyd

Lieferant: Apollo Scientific
Gefahr

PC0799-1G PC0799-250MG PC0799-5G
APOSPC0799-1GEA 741 EUR
APOSPC0799-1G APOSPC0799-250MG APOSPC0799-5G
4-(Pentafluorthio)benzaldehyd
4-(Pentafluorothio)benzaldehyde
Formel: C₇H₅F₅OS
Lagertemperatur: Raumtemperatur
MDL: MFCD03788518
CAS-Nummer: 401892-84-0
UN: 2810
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR