3-(Pentafluorthio)benzoesäure 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Carboxyphenylsulphur pentafluoride, 3-Sulphurpentafluorobenzoic acid

PC0800-1G PC0800-5G
APOSPC0800-1GEA 429 EUR
APOSPC0800-1G APOSPC0800-5G
3-(Pentafluorthio)benzoesäure 95%
3-(Pentafluorothio)benzoic acid
Formel: C₇H₅F₅O₂S
Molecular Weight: 248.17 g/mol
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788519
CAS-Nummer: 833-96-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR