Ethyl 2-cyano-3-(4-fluorophenyl)acrylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-cyano-3-(4-fluorophenyl)prop-2-enoate, Ethyl α-cyano-4-fluorocinnamate

PC0815-5G
APOSPC0815-5GEA 157 EUR
APOSPC0815-5G
Ethyl 2-cyano-3-(4-fluorophenyl)acrylate
Ethyl 2-cyano-3-(4-fluorophenyl)acrylate
Formel: C₁₂H₁₀FNO₂
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00176412
CAS-Nummer: 18861-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR