1-(Methylsulfonyl)-4-(trifluormethoxy)benzol 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(trifluoromethoxy)phenyl sulphone

PC0819-10G PC0819-5G
APOSPC0819-10GEA 343 EUR
APOSPC0819-10G APOSPC0819-5G
1-(Methylsulfonyl)-4-(trifluormethoxy)benzol 98%
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Formel: C₈H₇F₃O₃S
Schmelzpunkt: 64…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00792426
CAS-Nummer: 87750-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR