Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
PC0837-10G
PC0837-1G
APOSPC0837-10GEA
411
EUR
APOSPC0837-10G
APOSPC0837-1G
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Formel:
C₁₂H₁₂FN₃O₂ Schmelzpunkt: 153…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173917 CAS-Nummer: 138907-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...