Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

PC0837-10G PC0837-1G
APOSPC0837-10GEA 411 EUR
APOSPC0837-10G APOSPC0837-1G
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Formel: C₁₂H₁₂FN₃O₂
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173917
CAS-Nummer: 138907-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR