Allylheptafluoroisopropyl ether

Lieferant: Apollo Scientific
Achtung

PC0847-5G
APOSPC0847-5GEA 633 EUR
APOSPC0847-5G
Allylheptafluoroisopropyl ether
Allylheptafluoroisopropyl ether
Formel: C₆H₅F₇O
Molecular Weight: 226,09 g/mol
Siedepunkt: 64…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042148
CAS-Nummer: 15242-17-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR