6-Fluor-m-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC0854-25G
PC0854-5G
APOSPC0854-25GEA
0
EUR
APOSPC0854-25G
APOSPC0854-5G
6-Fluor-m-toluylsäure
6-Fluor-m-toluylsäure
Formel:
C₈H₇FO₂ Schmelzpunkt: 160…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092819 CAS-Nummer: 321-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...