2-Chlor-4-fluor-5-nitroanisol
Lieferant: Apollo Scientific
|
Achtung
|
PC0858-1G
PC0858-5G
PC0858-25G
APOSPC0858-1GEA
21.9
EUR
APOSPC0858-1G
APOSPC0858-5G
APOSPC0858-25G
2-Chlor-4-fluor-5-nitroanisol
2-Chlor-4-fluor-5-nitroanisol
Formel:
C₇H₅ClFNO₃ Schmelzpunkt: 72…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425810 CAS-Nummer: 84478-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...