(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
trans-3,4-(Difluoromethylenedioxy)cinnamic acid, (2E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)prop-2-enoic acid, 5-[(E)-2-Carboxyethenyl]-2,2-difluoro-1,3-benzodioxole
PC0880-1G
APOSPC0880-1GEA
226
EUR
APOSPC0880-1G
(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid 95%
(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid
Formel:
C₁₀H₆F₂O₄ Molecular Weight: 228,149 g/mol Schmelzpunkt: 176…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203111 CAS-Nummer: 387350-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...