(3-Fluor-2-methylphenyl)essigsäure 98%
Lieferant: Apollo Scientific
Synonyme:
2-(3-Fluoro-2-methylphenyl)acetic acid
PC0889-1G
PC0889-5G
PC0889-25G
APOSPC0889-1GEA
43.5
EUR
APOSPC0889-1G
APOSPC0889-5G
APOSPC0889-25G
(3-Fluor-2-methylphenyl)essigsäure 98%
(3-Fluor-2-methylphenyl)essigsäure
Formel:
C₉H₉FO₂ Molecular Weight: 168.17 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788545 CAS-Nummer: 500912-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...