2-Fluor-6-methoxybenzylbromid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Bromomethyl)-1-fluoro-3-methoxybenzene
PC0899-250MG
APOSPC0899-250MGEA
23.4
EUR
APOSPC0899-250MG
2-Fluor-6-methoxybenzylbromid
2-Fluor-6-methoxybenzylbromid
Formel:
C₈H₈BrFO Schmelzpunkt: 18…21 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08688579 CAS-Nummer: 500912-17-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...