4-Amino-2-chlor-5-nitrobenzotrifluorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-nitro-4-(trifluoromethyl)aniline
PC0912-5G
APOSPC0912-5GEA
75.4
EUR
APOSPC0912-5G
4-Amino-2-chlor-5-nitrobenzotrifluorid 97%
4-Amino-2-chlor-5-nitrobenzotrifluorid
Formel:
C₇H₄ClF₃N₂O₂ Schmelzpunkt: 113…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312277 CAS-Nummer: 35375-74-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...