(3-Trifluoromethylphenoxy)acetic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(3-(Trifluoromethyl)phenoxy)acetic acid

PC0923-1G
APOSPC0923-1GEA 132 EUR
APOSPC0923-1G
(3-Trifluoromethylphenoxy)acetic acid
(3-Trifluoromethylphenoxy)acetic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
Schmelzpunkt: 92…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03147381
CAS-Nummer: 349-82-6
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR