1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorophenyl)-1H-1,2,4-triazol-3-ol, 1-Fluoro-4-(3-hydroxy-1H-1,2,4-triazol-1-yl)benzene

PC0926-250MG
APOSPC0926-250MGEA 94.4 EUR
APOSPC0926-250MG
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
1-(4-Fluorophenyl)-3-hydroxy-1H-1,2,4-triazole
Formel: C₈H₆FN₃O
Molecular Weight: 179.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04035653
CAS-Nummer: 42158-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR