N-Boc-3-trifluoromethyl-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: Boc-Phe(3-CF3)-OH, (S)-2-((tert-Butoxycarbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid

PC0938-10G PC0938-1G PC0938-5G
APOSPC0938-10GEA 909 EUR
APOSPC0938-10G APOSPC0938-1G APOSPC0938-5G
N-Boc-3-trifluoromethyl-L-phenylalanine 97%
N-Boc-3-trifluoromethyl-L-phenylalanine
Formel: C₁₅H₁₈F₃NO₄
Schmelzpunkt: 135…138 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00672524
CAS-Nummer: 142995-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR