N-Boc-3-trifluoromethyl-L-phenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
Boc-Phe(3-CF3)-OH, (S)-2-((tert-Butoxycarbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid
PC0938-10G
PC0938-1G
PC0938-5G
APOSPC0938-10GEA
909
EUR
APOSPC0938-10G
APOSPC0938-1G
APOSPC0938-5G
N-Boc-3-trifluoromethyl-L-phenylalanine 97%
N-Boc-3-trifluoromethyl-L-phenylalanine
Formel:
C₁₅H₁₈F₃NO₄ Schmelzpunkt: 135…138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00672524 CAS-Nummer: 142995-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...