3-Fluoro-4-hydroxy-5-methoxybenzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-hydroxy-5-methoxybenzene

PC0941-1G
APOSPC0941-1GEA 65.4 EUR
APOSPC0941-1G
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
Formel: C₈H₇FO₃
Molecular Weight: 170,139 g/mol
Schmelzpunkt: 114…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683560
CAS-Nummer: 79418-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR