1-(3-(Trifluormethyl)phenyl)thioharnstoff 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-[3-(Trifluoromethyl)phenyl]thiourea
PC0942-25G
APOSPC0942-25GEA
147
EUR
APOSPC0942-25G
1-(3-(Trifluormethyl)phenyl)thioharnstoff 97%
1-(3-(Trifluormethyl)phenyl)thioharnstoff
Formel:
C₈H₇F₃N₂S Molecular Weight: 220,22 g/mol Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041171 CAS-Nummer: 1736-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...