Triethylammoniumdihydrogentrifluorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Triethylamin Trishydrofluorid, Triethylamin Trihydrofluorid
PC0945-500G
APOSPC0945-500GEA
176
EUR
APOSPC0945-500G
Triethylammoniumdihydrogentrifluorid
Triethylammonium dihydrogentrifluoride
Formel:
C₆H₁₈F₃N Molecular Weight: 161,21 g/mol Siedepunkt: 70 °C/15mmHg Schmelzpunkt: –29 °C Dichte: 0,989 g/cm³ (25 °C) Flammpunkt: 87 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00043294 CAS-Nummer: 73602-61-6 EINECS: 277-550-5 UN: 2927 ADR: 6.1+8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...