6-Fluorchromon
Lieferant: Apollo Scientific
Synonyme:
6-Fluoro-4H-chromen-4-one
PC0953-5G
APOSPC0953-5GEA
180
EUR
APOSPC0953-5G
6-Fluorchromon
6-Fluorchromon
Formel:
C₉H₅FO₂ Molecular Weight: 164,14 g/mol Schmelzpunkt: 166…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094002 CAS-Nummer: 105300-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...