6-Chloro-4-(trifluoromethyl)nicotinic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC0959-1G
PC0959-250MG
PC0959-5G
APOSPC0959-1GEA
156
EUR
APOSPC0959-1G
APOSPC0959-250MG
APOSPC0959-5G
6-Chloro-4-(trifluoromethyl)nicotinic acid 95+%
6-Chloro-4-(trifluoromethyl)nicotinic acid
Formel:
C₇H₃ClF₃NO₂ Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06800446 CAS-Nummer: 261635-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...