7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC0963-250MG
APOSPC0963-250MGEA
75.4
EUR
APOSPC0963-250MG
7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate 95+%
7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate
Formel:
C₁₀H₇FN₂O₃S Molecular Weight: 254,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445326 CAS-Nummer: 849924-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...