2-Fluoro-4-(methylsulphonyl)aniline 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC0964-25G
PC0964-5G
PC0964-100G
APOSPC0964-25GEA
0
EUR
APOSPC0964-25G
APOSPC0964-5G
APOSPC0964-100G
2-Fluoro-4-(methylsulphonyl)aniline 99%
2-Fluoro-4-(methylsulphonyl)aniline
Formel:
C₇H₈FNO₂S Schmelzpunkt: 145…147 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06800756 CAS-Nummer: 832755-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...