1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene

Lieferant: Apollo Scientific

PC0965-5G
APOSPC0965-5GEA 553 EUR
APOSPC0965-5G
1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene
1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene
Formel: C₃Cl₂F₄
Molecular Weight: 182,932 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09763641
CAS-Nummer: 2804-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR