1-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,4-diazepane

Lieferant: Apollo Scientific
Achtung

PC0966-1G PC0966-5G
APOSPC0966-1GEA 114 EUR
APOSPC0966-1G APOSPC0966-5G
1-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,4-diazepane
Formel: C₁₂H₁₇FN₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06800757
CAS-Nummer: 849924-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR