4-Brom-3,5-difluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC0968-5G
APOSPC0968-5GEA
21.9
EUR
APOSPC0968-5G
4-Brom-3,5-difluoranilin 97%
4-Brom-3,5-difluoranilin
Formel:
C₆H₄BrF₂N Molecular Weight: 208,01 g/mol Schmelzpunkt: 99…101 °C Flammpunkt: 89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236593 CAS-Nummer: 203302-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...