Perfluoro(3-methylbutan-2-one) 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0985-25G
PC0985-5G
APOSPC0985-25GEA
625
EUR
APOSPC0985-25G
APOSPC0985-5G
Perfluoro(3-methylbutan-2-one) 98%
Perfluoro(3-methylbutan-2-one)
Formel:
C₅F₁₀O Siedepunkt: 24 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784237 CAS-Nummer: 756-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...