Fluoroiodoacetic acid

Lieferant: Apollo Scientific
Gefahr

PC0992-1G PC0992-5G
APOSPC0992-1GEA 195 EUR
APOSPC0992-1G APOSPC0992-5G
Fluoroiodoacetic acid
Fluoroiodoacetic acid
Formel: C₂H₂FIO₂
Flammpunkt: 96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07784241
CAS-Nummer: 471-45-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR