1-Brom-2-fluor-4-nitrobenzol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brom-3-fluornitrobenzol

PC0996-25G PC0996-5G PC0996-100G
APOSPC0996-25GEA 42.2 EUR
APOSPC0996-25G APOSPC0996-5G APOSPC0996-100G
1-Brom-2-fluor-4-nitrobenzol 97%
1-Bromo-2-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Schmelzpunkt: 88…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865088
CAS-Nummer: 185331-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR