1-Brom-2-fluor-4-nitrobenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Brom-3-fluornitrobenzol
PC0996-25G
PC0996-5G
PC0996-100G
APOSPC0996-25GEA
42.2
EUR
APOSPC0996-25G
APOSPC0996-5G
APOSPC0996-100G
1-Brom-2-fluor-4-nitrobenzol 97%
1-Bromo-2-fluoro-4-nitrobenzene
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol Schmelzpunkt: 88…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05865088 CAS-Nummer: 185331-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...