3-Nitro-4-(trifluoromethyl)phenol 98%
Lieferant: Apollo Scientific
PC0997-1G
PC0997-5G
PC0997-10G
APOSPC0997-1GEA
0
EUR
APOSPC0997-1G
APOSPC0997-5G
APOSPC0997-10G
3-Nitro-4-(trifluoromethyl)phenol 98%
3-Nitro-4-(trifluoromethyl)phenol
Formel:
C7H4F3NO3 Schmelzpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07783660 CAS-Nummer: 25889-36-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...