4-Cyan-2-fluoranilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0999-25G
PC0999-5G
APOSPC0999-25GEA
27.7
EUR
APOSPC0999-25G
APOSPC0999-5G
4-Cyan-2-fluoranilin 98%
4-Cyan-2-fluoranilin
Formel:
C₇H₅FN₂ Molecular Weight: 136.13 g/mol Schmelzpunkt: 87…90 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00055559 CAS-Nummer: 63069-50-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...