N-Acetyl-2-fluoro-DL-phenylalanine 98%

Lieferant: Apollo Scientific

Synonyme: 2-(Acetylamino)-3-(2-fluorophenyl)propanoic acid

PC1000E-5G
APOSPC1000E-5GEA 80 EUR
APOSPC1000E-5G
N-Acetyl-2-fluoro-DL-phenylalanine 98%
N-Acetyl-2-fluoro-DL-phenylalanine
Formel: C₁₁H₁₂FNO₃
Molecular Weight: 225,216 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017915
CAS-Nummer: 66574-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR