Aflunox™ 606 95%

Lieferant: Apollo Scientific
Achtung

PC1002E-100G PC1002E-25G
APOSPC1002E-100GEA 56.7 EUR
APOSPC1002E-100G APOSPC1002E-25G
Aflunox™ 606 95%
Aflunox™

Aflunox oils are clear colourless oils suitable for use as lubricants and are inert to oxidising agents such as fluorine, chlorine and oxygen at high temperatures.


  • F[CF(CF3)CF2O]xC2F5 x = 12
  • viscosity 68 cst@20C


Aflunox is a registered trademark of Japan Mektron

Schmelzpunkt: –60 °C
Dichte: 1.88 g/cm3 @ 25°C
MDL: MFCD00166448
CAS-Nummer: 52700-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR