Aflunox™ 1406

Lieferant: Apollo Scientific

PC1002G-100G
APOSPC1002G-100GEA 379 EUR
APOSPC1002G-100G
Aflunox™ 1406
Aflunox™

Aflunox oils are clear colourless oils suitable for use as lubricants and are inert to oxidising agents such as fluorine, chlorine and oxygen at high temperatures.


Aflunox is a registered trademark of Japan Mektron

Schmelzpunkt: –55 °C
Dichte: 1.89 g/cm3 @ 25°C
MDL: MFCD00166448
CAS-Nummer: 52700-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR