Aflunox™ 2507

Lieferant: Apollo Scientific
Achtung

PC1002N-100G PC1002N-25G
APOSPC1002N-100GEA 62.5 EUR
APOSPC1002N-100G APOSPC1002N-25G
Aflunox™ 2507
Aflunox™

Perfluoropolyether fluids are inert to aggressive reagents including elemental fluorine. Aflunox 2507 is an excellent inert lubricant for laboratory stirrer seals.


  • F[CF(CF3)CF2O]xC2F5 x = 21
  • viscosity 270 cSt@20C


Aflunox is a registered trademark of Japan Mektron

Schmelzpunkt: –45 °C
Dichte: 1.89 g/cm3 @ 25°C
MDL: MFCD00166448
CAS-Nummer: 52700-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR