Aflunox™ 2507
Lieferant: Apollo Scientific
|
Achtung
|
PC1002N-100G
PC1002N-25G
APOSPC1002N-100GEA
62.5
EUR
APOSPC1002N-100G
APOSPC1002N-25G
Aflunox™ 2507
Aflunox™
Perfluoropolyether fluids are inert to aggressive reagents including elemental fluorine. Aflunox 2507 is an excellent inert lubricant for laboratory stirrer seals.
- F[CF(CF3)CF2O]xC2F5 x = 21
- viscosity 270 cSt@20C
Aflunox is a registered trademark of Japan Mektron
Schmelzpunkt:
–45 °C Dichte: 1.89 g/cm3 @ 25°C |
MDL:
MFCD00166448 CAS-Nummer: 52700-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...