2,3,5,6-Tetrafluor-p-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC10050-1G
APOSPC10050-1GEA
109
EUR
APOSPC10050-1G
2,3,5,6-Tetrafluor-p-toluylsäure
2,3,5,6-Tetrafluor-p-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208,11 g/mol Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012302 CAS-Nummer: 652-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...