4-Allyl-2,3,5,6-tetrafluorobenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC1006T-250MG
APOSPC1006T-250MGEA 21.9 EUR
APOSPC1006T-250MG
4-Allyl-2,3,5,6-tetrafluorobenzoic acid 97%
4-Allyl-2,3,5,6-tetrafluorobenzoic acid
Formel: C₁₀H₆F₄O₂
Schmelzpunkt: 89…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082610
CAS-Nummer: 79538-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR