3,5-Dimethyl-2,4,6-trifluorobenzoyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-(Chloroformyl)-2,4,6-trifluoro-m-xylene, 5-(Chlorocarbonyl)-2,4,6-trifluoro-m-xylene
PC10084-1G
APOSPC10084-1GEA
232
EUR
APOSPC10084-1G
3,5-Dimethyl-2,4,6-trifluorobenzoyl chloride
3,5-Dimethyl-2,4,6-trifluorobenzoyl chloride
Formel:
C₉H₆ClF₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039201 CAS-Nummer: 886762-25-0 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...