2-Fluor-3-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC10096-100G
PC10096-25G
PC10096-5G
APOSPC10096-100GEA
319
EUR
APOSPC10096-100G
APOSPC10096-25G
APOSPC10096-5G
2-Fluor-3-methoxyphenylboronsäure
2-Fluor-3-methoxyphenylboronsäure
Formel:
C₇H₈BFO₃ Schmelzpunkt: 117…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094728 CAS-Nummer: 352303-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...